MEM, no glutamine, no phenol red, 10 x 500 mL - FAQs
MEM, no glutamine, no phenol red, 10 x 500 mL - FAQs
View additional product information for MEM, no glutamine, no phenol red - FAQs (51200087)
2 product FAQs found
Where can I find the osmolality for MEM Medium?
The osmolality is listed in the COA for the particular lot number of the medium.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
Do you offer media without phenol red (phenol red-free media)?
Yes, please visit this page for a list of our phenol red-free media (http://tools.thermofisher.com/content/sfs/brochures/Phenol_RedFree_Media_Glance.pdf).
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.