NFkB p50 Polyclonal Antibody - FAQs
NFkB p50 Polyclonal Antibody - FAQs
View additional product information for NFkB p50 Polyclonal Antibody - FAQs (513500)
1 product FAQs found
Can NFkB p50 Polyclonal Antibody (Cat. No. 513500) detect both precursor protein p105 and its cleavage product p50?
Sorry, we have not analyzed this.