D.A.R.T.'S™ (Disposable Automation Research Tips)
Actual product may vary
Thermo Scientific™
D.A.R.T.'S™ (Disposable Automation Research Tips)
Achieve ultimate performance in automated pipetting with these tips--superior quality with the flexibility to choose tips for any application.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)SterilityFilteredPackaging
531130 μLNon-sterileNon-filtered10 x 384 tips
530112.5 μLNon-sterileNon-filtered10 x 384 tips
5417R30 μLSterileNon-filtered
530212.5 μLSterileNon-filtered10 x 384 tips
531730 μLSterileNon-filtered20 x 384 tips
531830 μLSterileFiltered20 x 384 tips
5321100 μLNon-sterileNon-filtered10 x 384 tips
5326100 μLNon-sterileNon-filtered20 x 384 tips
5327100 μLSterileNon-filtered20 x 384 tips
5328100 μLSterileFiltered20 x 384 tips
5516TS
also known as 5516
300 μLNon-sterileNon-filtered20 x 96 tips
5517TS
also known as 5517
300 μLSterileNon-filtered20 x 96 tips
5518300 μLSterileFiltered20 x 96 tips
5526100 μLNon-sterileNon-filtered20 x 96 tips
5527100 μLSterileNon-filtered20 x 96 tips
5528100 μLSterileFiltered20 x 96 tips
558630 μLNon-sterileNon-filtered20 x 96 tips
558730 μLSterileNon-filtered20 x 96 tips
558830 μLSterileFiltered20 x 96 tips
5542250 μLSterileNon-filtered20 x 96 tips
5545220 μLSterileFiltered20 x 96 tips
557130 μLNon-sterileNon-filtered20 x 96 tips
5322100 μLSterile10 x 384 tips
5572TS
also known as 5572
30 μLSterileNon-filtered20 x 96 tips
531630 μLNon-sterileNon-filtered20 x 384 tips
Catalog number 5311
Price (USD)
1,045.50
Case of 3840
Volume (Metric):
30 μL
Sterility:
Non-sterile
Filtered:
Non-filtered
Packaging:
10 x 384 tips
Price (USD)
1,045.50
Case of 3840
Perform pipetting more effectively with Thermo Scientific™ Matrix™ SerialMate™ Pipette Tips, for use with the Matrix SerialMate™ Pipetter. Unlike conventional automation tips, D.A.R.T.'s™ are packaged in disposable rigid magazines and seal onto the pipetting heads by being pressed against a silicone pad. This definitive seal minimizes volume displacement, ensures tip straightness and enables consistent dispensing for improved well access. A small internal orifice provides small droplet formation resulting in optimal accuracy and precision.
Features:
  • Stepped-body design minimizes volume displacement and ensures tip straightness for better well access.
  • Highly polished molds and cores ensure low liquid retention.
  • Small internal tip orifice provides better small volume droplet formation
  • Sterile, sterile filtered, wide-bore, and extended-length tip options are available
  • Tips are thoroughly inspected for proper formation and straightness
  • Unique face seal design ensures consistent tip-to-tip height for precise low volume dispensing.
  • Entire assembly can be quickly removed and replaced eliminating extra handling of the tips, which can cause damage or contamination.
Includes: 20 magazines per case
Compatible with: Thermo Scientific™ Matrix™ Hydra™ DT, Matrix™ PlateMate™ 2x3 and PlateMate™ Plus Systems.
Specifications
Certifications/ComplianceISO 9001 and ISO 13485
Clean ClaimsFree of RNase, DNase, DNA, ATP, and endotoxin
Description384 Tips, Requires special aluminum tip magazine
DisposableSingle-use
Packaging10 x 384 tips
Quantity3840 tips
AutoclavableAutoclavable
ColorNatural
FilteredNon-filtered
For Use With (Equipment)Thermo Scientific Matrix SerialMate
FormatRack
Product LineMatrix SerialMate
SterilityNon-sterile
Tip StyleAutomation
Volume (Metric)30 μL
Unit SizeCase of 3840