Pierce™ Streptavidin Plus UltraLink™ Resin
Thermo Scientific™

Pierce™ Streptavidin Plus UltraLink™ Resin

The Thermo Scientific™ Pierce™ High Capacity Streptavidin Chromatography Cartridges are convenient, ready-to-use devices for separating biotinylated molecules from non-biotinylated moleculesRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
531175 mL
531162 mL
Catalog number 53117
Price (USD)
796.00
Each
Quantity:
5 mL
Request bulk or custom format
Price (USD)
796.00
Each
The Thermo Scientific™ Pierce™ High Capacity Streptavidin Chromatography Cartridges are convenient, ready-to-use devices for separating biotinylated molecules from non-biotinylated molecules and for purifying antigens using biotinylated antibodies. The chromatography cartridges contain a beaded resin of immobilized recombinant streptavidin protein that binds biotinylated proteins at greater than 10 mg/mL of beads. This corresponds to 2 to 3 times higher biotin-binding capacity than products from other suppliers and is ideal when a high amount of recovery is required.

These cartridges enable fast, easy, and reproducible chromatographic separations and are compatible with the major automated liquid-chromatography systems or manual syringe processing (see Table 1 for cartridge general properties). The cartridges attach directly to ÄKTA™ or FPLC systems without additional connectors. The included accessory pack readily adapts cartridges for use with Luer-Lok™ syringe fittings or 1/16' tubing. These cartridges can be regenerated multiple times for affinity purification.

Features of High Capacity Streptavidin Agarose:
• Streptavidin—purified recombinant streptavidin protein (53 kDa, near-neutral pI); tetrameric with four biotin-binding site per molecule
• Agarose resin—support is cross-linked 6% beaded agarose (CL-6B), the most popular resin for protein affinity purification methods
• Inert and stable—streptavidin is immobilized by charge-free, leach-resistant covalent bonds, resulting in low nonspecific binding and enabling multiple uses without decline in yield
• High capacity—this variety of beads has a dense load of immobilized streptavidin, providing a binding capacity greater than 10 mg of biotinylated BSA per mL of resin
• Two formats—available in several bottle sizes of resin slurry and as packed 1 mL and 5 mL chromatography cartridges

Properties of cross-linked 6% beaded agarose (CL-6B):
• Support pH stability: 2 to 14 (short term); 3 to 13 (long term)
• Average particle size: 45 to 165 microns
• Exclusion limit: 10,000 to 4,000,000 daltons
• Maximum recommended flow rate: 4 mL/minute (1 mL column), 5 mL/min (5 mL column)
• Maximum linear velocity: 30 cm per hour
• Maximum pressure: 0.3 MPa, 43.5 psi or 3 bar

Pierce High Capacity Streptavidin Chromatography Cartridges consist of purified recombinant streptavidin covalently immobilized at a high density onto high-quality cross-linked 6% beaded agarose. Biotinylated antibodies, proteins, peptides, nucleic acids, and other molecules or interaction complexes can be captured, immunoprecipitated, removed, or purified from samples using this streptavidin resin.

Streptavidin is a tetrameric protein containing four biotin-binding sites similar to avidin. The native protein from Streptomyces avidinii is carbohydrate-free, has an acidic pI of 5.5 and a mass of 75 kDa. The recombinant protein used to manufacture Pierce streptavidin resins has a near-neutral isoelectric point and a mass of 53 kDa. Streptavidin generally has less nonspecific binding than avidin (from chicken egg white) because of the absence of carbohydrates.

For more information see: Chromatography Cartridges for Desalting and Affinity Purification
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Column TypeAffinity, UltraLink™ Resin
FormatBottle
Protein FormRecombinant
Quantity5 mL
Stationary PhaseStreptavidin
Sufficient ForBinding ≥/= 48 μg Biotin/mL Resin (≥/= 4 mg Biotinylated BSA/mL Resin)
TargetBiotin-Tagged Molecule
Capacity (Metric)10 mg Biotinylated BSA Binding per mL Resin
Particle Size45 to 165 μm
Product LinePierce, UltraLink
TypeResin
Unit SizeEach
Contents & Storage
Upon receipt store at 4°C. Product is shipped at ambient temperature.

Frequently asked questions (FAQs)

When should the UltraLink Immobilized Streptavidin Plus Support be used in place of streptavidin immobilized on agarose?

UltraLink Immobilized Streptavidin Plus Support should be used when you are planning to perform anything other than gravity filtration with a column. Streptavidin Plus UltraLink Resin is often preferred for immunoprecipitation because the white appearance of the support allows the pellet to be visualized more easily than the translucent pellet of agarose supports.

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.

What is the biotin-binding capacity for UltraLink Biosupport Medium?

The biotin-binding capacity is >3 mg biotinylated BSA/ml of resin, or >120 nMoles biotin/mL of resin which is equilvalent to approximately 30 µg of biotin or 30 units of biotin/ml.

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.

What is the affinity of streptavidin for biotin in the Thermo Scientific Streptavidin Plus Ultralink Resin kit?

The binding affinity of streptavidin is similar to that of avidin, which is Ka = 10^15/M.

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.

What is UltraLink Biosupport Medium?

UltraLink Biosupport Medium is stronger than agarose, being capable of withstanding 100 p.s.i. The support starts with an azlactone ring that opens upon immobilization of a ligand to form a five-molecule spacer arm.

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.

What can prevent biotin from binding to the immobilized streptavidin when using the Thermo Scientific Streptavidin Plus Ultralink Resin?

Steric hindrance can interfere with the binding. As with avidin, the double ring structure of biotin must fit 9 angstroms into the streptavidin molecule for binding to occur. Competitive inhibition by endogenous biotin may also interfere with binding of your biotinylated ligand.

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.