D.A.R.T.'S™ (Disposable Automation Research Tips)
Actual product may vary
Thermo Scientific™
D.A.R.T.'S™ (Disposable Automation Research Tips)
Achieve ultimate performance in automated pipetting with these tips--superior quality with the flexibility to choose tips for any application.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)SterilityFilteredPackaging
531730 μLSterileNon-filtered20 x 384 tips
530112.5 μLNon-sterileNon-filtered10 x 384 tips
5417R30 μLSterileNon-filtered
530212.5 μLSterileNon-filtered10 x 384 tips
531830 μLSterileFiltered20 x 384 tips
5321100 μLNon-sterileNon-filtered10 x 384 tips
5326100 μLNon-sterileNon-filtered20 x 384 tips
5327100 μLSterileNon-filtered20 x 384 tips
5328100 μLSterileFiltered20 x 384 tips
5516TS
also known as 5516
300 μLNon-sterileNon-filtered20 x 96 tips
5517TS
also known as 5517
300 μLSterileNon-filtered20 x 96 tips
5518300 μLSterileFiltered20 x 96 tips
5526100 μLNon-sterileNon-filtered20 x 96 tips
5527100 μLSterileNon-filtered20 x 96 tips
5528100 μLSterileFiltered20 x 96 tips
558630 μLNon-sterileNon-filtered20 x 96 tips
558730 μLSterileNon-filtered20 x 96 tips
558830 μLSterileFiltered20 x 96 tips
531130 μLNon-sterileNon-filtered10 x 384 tips
5542250 μLSterileNon-filtered20 x 96 tips
5545220 μLSterileFiltered20 x 96 tips
557130 μLNon-sterileNon-filtered20 x 96 tips
5322100 μLSterile10 x 384 tips
5572TS
also known as 5572
30 μLSterileNon-filtered20 x 96 tips
531630 μLNon-sterileNon-filtered20 x 384 tips
Catalog number 5317
Price (USD)
1,680.24
Online exclusive
2,133.53
Save 453.29
Case of 7680
Volume (Metric):
30 μL
Sterility:
Sterile
Filtered:
Non-filtered
Packaging:
20 x 384 tips
Price (USD)
1,680.24
Online exclusive
2,133.53
Save 453.29
Case of 7680
Perform pipetting more effectively with Thermo Scientific™ Matrix™ SerialMate™ Pipette Tips, for use with the Matrix SerialMate™ Pipetter. Unlike conventional automation tips, D.A.R.T.'s™ are packaged in disposable rigid magazines and seal onto the pipetting heads by being pressed against a silicone pad. This definitive seal minimizes volume displacement, ensures tip straightness and enables consistent dispensing for improved well access. A small internal orifice provides small droplet formation resulting in optimal accuracy and precision.
Features:
  • Stepped-body design minimizes volume displacement and ensures tip straightness for better well access.
  • Highly polished molds and cores ensure low liquid retention.
  • Small internal tip orifice provides better small volume droplet formation
  • Sterile, sterile filtered, wide-bore, and extended-length tip options are available
  • Tips are thoroughly inspected for proper formation and straightness
  • Unique face seal design ensures consistent tip-to-tip height for precise low volume dispensing.
  • Entire assembly can be quickly removed and replaced eliminating extra handling of the tips, which can cause damage or contamination.
Includes: 20 magazines per case
Compatible with: Thermo Scientific™ Matrix™ Hydra™ DT, Matrix™ PlateMate™ 2x3 and PlateMate™ Plus Systems.
Specifications
Certifications/ComplianceISO 9001 and ISO 13485
Clean ClaimsFree of RNase, DNase, DNA, ATP, and endotoxin
DisposableSingle-use
Length (English)Standard
Packaging20 x 384 tips
Quantity7680 tips
AutoclavableNot Autoclavable
ColorNatural
FilteredNon-filtered
For Use With (Equipment)Thermo Scientific Matrix Hydra DT, Thermo Scientific PlateMate Plus, Thermo Scientific Versette
FormatRack
Product LineD.A.R.T.S
SterilitySterile
Tip StyleAutomation
Volume (Metric)30 μL
Unit SizeCase of 7680
Frequently asked questions (FAQs)
How are the D.A.R.T. tips packaged for the Versette Automated Liquid Handler?
The D.A.R.T. tips for the Versette Automated Liquid Handler are packaged as shown below:
Each case contains 4 sleeves and each sleeve in turn contains 5 tip magazines. Hence, each case has 20 tip magazines.
- For the 96-channel pipetting head, each case contains 96 x 20 = 1,920 tips
- For the 384-channel pipetting head, each case contains 384x 20 = 7,680 tips
Can I use any pipette tips with the Versette Automated Liquid Handler?
The Versette Automated Liquid Handler uses a closed-tip platform incorporating D.A.R.T. (Disposable Automation Research Tips) tips. D.A.R.T.s are in disposable rigid magazines and seal onto the pipetting heads by being pressed against a silicone pad. They help ensure accurate and precise pipetting across all channels and are designed to deliver a “no-leak” seal every time.