D.A.R.T.'S™ (Disposable Automation Research Tips) - FAQs
D.A.R.T.'S™ (Disposable Automation Research Tips) - FAQs
View additional product information for D.A.R.T.'S™ (Disposable Automation Research Tips) - FAQs (5321, 5586, 5542, 5322, 5301, 5587, 5572TS, 5316, 5317, 5318, 5518, 5571, 5311, 5417R, 5516TS, 5517TS, 5302, 5588, 5545, 5326, 5327, 5328, 5526, 5527, 5528)
2 product FAQs found
How are the D.A.R.T. tips packaged for the Versette Automated Liquid Handler?
The D.A.R.T. tips for the Versette Automated Liquid Handler are packaged as shown below:
Each case contains 4 sleeves and each sleeve in turn contains 5 tip magazines. Hence, each case has 20 tip magazines.
- For the 96-channel pipetting head, each case contains 96 x 20 = 1,920 tips
- For the 384-channel pipetting head, each case contains 384x 20 = 7,680 tips
Can I use any pipette tips with the Versette Automated Liquid Handler?
The Versette Automated Liquid Handler uses a closed-tip platform incorporating D.A.R.T. (Disposable Automation Research Tips) tips. D.A.R.T.s are in disposable rigid magazines and seal onto the pipetting heads by being pressed against a silicone pad. They help ensure accurate and precise pipetting across all channels and are designed to deliver a “no-leak” seal every time.