Product Image
Applied Biosystems™

Pin, Needle Valve

This is a spare part for the 310 Genetic Analyzer.
Have Questions?
Catalog NumberQuantity
53991 unit
Catalog number 5399
Price (USD)
297.00
Each
Quantity:
1 unit
Price (USD)
297.00
Each

This is a spare part for the 310 Genetic Analyzer.

For Research Use Only. Not for use in diagnostic procedures.

Specifications
For Use With (Equipment)310 Genetic Analyzer
Product TypeNeedle Valve Pin
Quantity1 unit
Shipping ConditionRoom Temperature
Unit SizeEach
Contents & Storage
Store at room temperature.