KingFisher™ Purification System - FAQs
KingFisher™ Purification System - FAQs
View additional product information for KingFisher™ Purification System - FAQs (5400000)
1 product FAQs found
What is BindIt Software?
BindIt Software is used to operate the KingFisher 96, KingFisher mL, KingFisher Flex, KingFisher Duo Prime, and KingFisher Presto instruments. With the software you can:
• Create a plate layout
• Create new or modify existing protocols
• Transfer protocols to the KingFisher instrument memory
• Delete protocols from the KingFisher instrument
• Execute a protocol directly from the software without first sending it to the KingFisher instrument memory
• Export protocols to other databases
• Import protocols to the PC
• View protocol status reports