Do you have scripts for using the MagMAX kits on the KingFisher mL instrument?
We do not have prewritten scripts for using MagMAX kits on the KingFisher mL instrument. We recommend adapting scripts written for the KingFisher Flex or KingFisher Presto instruments. You will still need to validate the scripts on your KingFisher mL instrument.
Find additional tips, troubleshooting help, and resources within our Automated Nucleic Acid Purification Support Center.
Find additional tips, troubleshooting help, and resources within our Automated Nucleic Acid Purification Support Center.