Product Image
Applied Biosystems™

Syringe Ferrule, Bushing Seal (for the 310 Genetic Analyzer)

This is a component of the glass syringes used with the 310 Genetic Analyzer.
Have Questions?
Catalog NumberQuantity
54011 each
Catalog number 5401
Price (USD)
320.00
Each
Quantity:
1 each
Price (USD)
320.00
Each

This is a component of the glass syringes used with the 310 Genetic Analyzer.

For Research Use Only. Not for use in diagnostic procedures.

Specifications
For Use With (Equipment)310 Genetic Analyzer
Product TypeSyringe Ferrule
Quantity1 each
Shipping ConditionRoom Temperature
Unit SizeEach
Contents & Storage
Store at room temperature.