Product Image
Applied Biosystems™

Anode Buffer Reservoir Jar

This Buffer Reservoir is for use with the 3100-Avant™ Genetic Analyzer.
Have Questions?
Catalog NumberQuantity
54021 jar
Catalog number 5402
Price (USD)
221.00
Each
Quantity:
1 jar
Price (USD)
221.00
Each

This Buffer Reservoir is for use with the 3100-Avant™ Genetic Analyzer. It attaches to the lower polymer block, holding the anodic buffer.

For Research Use Only. Not for use in diagnostic procedures.

Specifications
For Use With (Application)Sequencing
For Use With (Equipment)310 Genetic Analyzer
No. of Reactions13
Product TypeAnode Buffer Reservoir Jar
Quantity1 jar
Shipping ConditionRoom Temperature
Unit SizeEach
Contents & Storage
Store at room temperature.