D.A.R.T.'S™ (Disposable Automation Research Tips), with Extended Length
Actual product may vary
Thermo Scientific™
D.A.R.T.'S™ (Disposable Automation Research Tips), with Extended Length
Achieve ultimate performance in automated pipetting with Thermo Scientific™ D.A.R.T.'S™ Extended Length tips — superior quality with the flexibility to choose tips for any application.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)SterilityFilteredTip Style
541630 μLNon-sterileNon-filteredLow Retention, Extended Length
541830 μLNon-sterileFilteredLow Retention, Extended Length
5538300 μLSterileFilteredLow Retention, Extended Length
5548300 μLSterileFilteredLow Retention, Wide Bore, Extended Length
5561220 μLNon-sterileNon-filteredExtended Length
550630 μLNon-sterileNon-filteredLow Retention, Extended Length
5536300 μLNon-sterileNon-filteredLow Retention, Extended Length
5546TS
also known as 5546
300 μLNon-sterileNon-filteredLow Retention, Wide Bore, Extended Length
541730 μLSterileNon-filteredLow Retention, Extended Length
550730 μLSterileNon-filteredLow Retention, Extended Length
5537300 μLSterileNon-filteredLow Retention, Extended Length
5547300 μLSterileNon-filteredLow Retention, Wide Bore, Extended Length
Catalog number 5416
Price (USD)
1,746.63
Online exclusive
2,081.77
Save 335.14
Case of 7680
Volume (Metric):
30 μL
Sterility:
Non-sterile
Filtered:
Non-filtered
Tip Style:
Low Retention, Extended Length
Price (USD)
1,746.63
Online exclusive
2,081.77
Save 335.14
Case of 7680
Perform pipetting more effectively with Thermo Scientific™ Matrix™ SerialMate™ Extended Length Pipette Tips, for use with the Matrix SerialMate™ Pipetter. Unlike conventional automation tips, D.A.R.T.'s™ are packaged in disposable rigid magazines and seal onto the pipetting heads by being pressed against a silicone pad. This definitive seal minimizes volume displacement, ensures tip straightness and enables consistent dispensing for improved well access. A small internal orifice provides small droplet formation resulting in optimal accuracy and precision.
Features:
  • Stepped-body design minimizes volume displacement and ensures tip straightness for better well access.
  • Face seal design ensures consistent tip-to-tip height for precise low volume dispensing
  • Highly polished molds and cores ensure low liquid retention.
  • Small internal tip orifice provides better small volume droplet formation, resulting in optimal accuracy and precision.
  • Sterile, sterile filtered and wide-bore tip options are available
  • Tips are thoroughly inspected for proper formation and straightness
  • Unique face seal design ensures consistent tip-to-tip height for precise low volume dispensing.
  • Entire assembly can be quickly removed and replaced eliminating extra handling of the tips, which can cause damage or contamination.
Includes: 20 magazines per case
Compatible with: Thermo Scientific™ Matrix™ Hydra™ DT, Matrix™ PlateMate™ 2x2, Matrix™ PlateMate™ 2x3, PlateMate™ Plus Systems and Versette.
Specifications
Certifications/ComplianceISO 9001 and ISO 13485
Clean ClaimsFree of RNase, DNase, DNA, ATP, and endotoxin
DisposableSingle-use
Packaging20 x 384 Tips
Quantity7680 tips
AutoclavableAutoclavable
ColorNatural
FilteredNon-filtered
For Use With (Equipment)Thermo Scientific Matrix Hydra DT, Thermo Scientific PlateMate Plus, Thermo Scientific Versette
Product LineMatrix
SterilityNon-sterile
Tip StyleLow Retention, Extended Length
Volume (Metric)30 μL
Unit SizeCase of 7680
Frequently asked questions (FAQs)
How are the D.A.R.T. tips packaged for the Versette Automated Liquid Handler?
The D.A.R.T. tips for the Versette Automated Liquid Handler are packaged as shown below:
Each case contains 4 sleeves and each sleeve in turn contains 5 tip magazines. Hence, each case has 20 tip magazines.
- For the 96-channel pipetting head, each case contains 96 x 20 = 1,920 tips
- For the 384-channel pipetting head, each case contains 384x 20 = 7,680 tips
Can I use any pipette tips with the Versette Automated Liquid Handler?
The Versette Automated Liquid Handler uses a closed-tip platform incorporating D.A.R.T. (Disposable Automation Research Tips) tips. D.A.R.T.s are in disposable rigid magazines and seal onto the pipetting heads by being pressed against a silicone pad. They help ensure accurate and precise pipetting across all channels and are designed to deliver a “no-leak” seal every time.