D.A.R.T.'S™ (Disposable Automation Research Tips), with Extended Length - FAQs
D.A.R.T.'S™ (Disposable Automation Research Tips), with Extended Length - FAQs
View additional product information for D.A.R.T.'S™ (Disposable Automation Research Tips), with Extended Length - FAQs (5561, 5546TS, 5536, 5547, 5548, 5416, 5537, 5538, 5417, 5418, 5506, 5507)
2 product FAQs found
How are the D.A.R.T. tips packaged for the Versette Automated Liquid Handler?
The D.A.R.T. tips for the Versette Automated Liquid Handler are packaged as shown below:
Each case contains 4 sleeves and each sleeve in turn contains 5 tip magazines. Hence, each case has 20 tip magazines.
- For the 96-channel pipetting head, each case contains 96 x 20 = 1,920 tips
- For the 384-channel pipetting head, each case contains 384x 20 = 7,680 tips
Can I use any pipette tips with the Versette Automated Liquid Handler?
The Versette Automated Liquid Handler uses a closed-tip platform incorporating D.A.R.T. (Disposable Automation Research Tips) tips. D.A.R.T.s are in disposable rigid magazines and seal onto the pipetting heads by being pressed against a silicone pad. They help ensure accurate and precise pipetting across all channels and are designed to deliver a “no-leak” seal every time.