Axiom™ Buffalo Genotyping Array
Applied Biosystems™

Axiom™ Buffalo Genotyping Array

Axiom Buffalo Genotyping Array was designed by the Affymetrix Expert Design Program.* The array was developed in collaboration with theRead more
Have Questions?
Catalog NumberQuantity
5504311 plate
Catalog number 550431
Price (USD)
-
Quantity:
1 plate
Axiom Buffalo Genotyping Array was designed by the Affymetrix Expert Design Program.* The array was developed in collaboration with the International Buffalo Genome Consortia, which included Fondazione Parco Tecnologico Padano in Italy, Iowa State University, and the Agricultural Research Service, United States Department of Agriculture (USDA).

Axiom Buffalo Genotyping Array provides the highest genome-wide coverage of polymorphic SNPs across multiple species and breeds of the water buffalo (Bubalus bubalis bubalis and Bubalus bubalis carabanensis), one of the most important beef and dairy animals in Italy, Brazil, and South Asia.

This array is the only commercially available high-density buffalo genotyping tool.

Features of Axiom Buffalo Genotyping Array:
• Comprehensive content – 90,000 common and rare markers selected from sequencing initiatives of the water buffalo (B. bubalis bubalis)
• Multiple breeds represented – Mediterranean, Murrah, Jaffarabadi, and Nili-Ravi
• Robust assay performance – SNPs demonstrate a ≥99%; call rate and ≥80% conversion rate on Murrah and Mediterranean samples

Applications of Axiom Buffalo Genotyping Array:
• Molecular breeding:
   - Genomic biomarker discovery
   - Marker-assisted selection
   - Routine screening
   - DNA fingerprinting for animal tracking

• Complex trait research:
   - High-resolution mapping of genetic loci in complex traits such as milk production, percentage of milk fat, and feed conversion efficiency
   - Discovery of genes and pathways underlying simple and complex traits

• Conservation and biodiversity:
   - Population diversity studies in river and swamp buffalo
   - Genomic marker-assisted conservation strategies

*Expert Design Arrays are designed by key opinion leaders in the plant and animal genomics community and made available to the public for the purpose of advancing research and breeding programs. These arrays contain genetic markers and annotations that are provided by the original customer who designed the array and are therefore not updated in the same manner as other catalog products. Through the program, we continually provides agrigenomics researchers access to genotyping arrays that have been utilized by their peers in the research community.

Required Products
Axiom Analysis Suite
GeneChip Command Console Software
GeneTitan Multi-Channel Instrument
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)Microarray Analysis
Quantity1 plate
ArrayGenotyping
Format96-array Plate
Number of Arrays96 arrays
SpeciesBuffalo
Unit Size1 plate

Frequently asked questions (FAQs)

What is the starting amount of DNA required for Axiom arrays?

All Axiom arrays (except for Axiom Genome-Wide Pan-African Array Set) require a total of 200 ng DNA. The Axiom Genome-Wide Pan-African Array Set requires a total of 300 ng DNA, or 100 ng DNA per array (there are 3 arrays).

Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.

What are the sample requirements for Axiom array processing at the Thermo Fisher Scientific Microarray Research Services Laboratory (MRSL)?

For information regarding our in-house MRSL requirements, please contact us via phone at 888-362-2447 (in North America) or via email at BioinformaticsServices@thermofisher.com.

Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.