Axiom™ Equine Genotyping Array
Applied Biosystems™
Axiom™ Equine Genotyping Array
Axiom Equine Genotyping Array (Axiom MNEC670) for equine genotyping was designed through the Expert Design Program by Affymetrix in collaboration詳細を見る
製品番号(カタログ番号)数量
5505831 plate
製品番号(カタログ番号) 550583
価格(JPY)
-
数量:
1 plate
Axiom Equine Genotyping Array (Axiom MNEC670) for equine genotyping was designed through the Expert Design Program by Affymetrix in collaboration with the University of Minnesota and members of the Equine Genetic Diversity Consortium.*
Axiom Equine Genotyping Array is a novel array and the only array that can be used for genotyping 20 different breeds. It offers maximum utility for research in equine genetics. It features 670,796 markers that were selected through a screening of 2 million markers for optimal genomic coverage of known genetic diversity among domestic horse breeds.1 Powered by Axiom Genotyping Solution, the array offers the highest data completeness, call rates, and reproducibility of any technology available today.
Features of Axiom™ Equine Genotyping Array:
• Expert design: developed by key opinion leaders in the equine community
• High resolution: available in 96 format; supporting a wide range of genotyping applications for equine research
Highlights
Content:
• Axiom Equine Genotyping Array includes a total of 670,796 markers:
   - Approximately 400,000 markers between population tag SNPs
   - Approximately 200,000 markers within population tag SNPs
   - Approximately 70,000 SNPs from legacy arrays
   - Additional SNP density within the equine major histocompatibility complex
Diversity:
Axiom Equine Genotyping Array includes markers that were selected to maximize accuracy of imputation up to 1.8 million markers both within and across 20 breeds(Fig. 1). An emphasis was placed on breeds with high commercial relevance.
Applications of Axiom Equine Genotyping Array:
• Complex trait research
   - Genome-wide analysis and fine mapping
   - Diversity analyses
   - Genomic prediction of disease risk
• Molecular breeding
   - Association mapping
   - Improved efficiency of breeding programs
   - Selection signature analyses
*The Equine Genetic Diversity Consortium (EGDC) led by the University of Minnesota represents a collaborative international community of equine researchers who are working to build a comprehensive understanding of genetic diversity among equine populations across the world. (http://www.cvm.umn.edu/equinegenetics/egdc/)
Required Products
GeneChip Command Console Software
GeneTitan Multi-Channel Instrument
研究用途にのみご使用ください。診断目的には使用できません。
仕様
使用対象(アプリケーション)Microarray Analysis
数量1 plate
アレイGenotyping
フォーマット96-array Plate
アレイの数96 arrays
Equine
Unit Size1 plate
よくあるご質問(FAQ)
What is the starting amount of DNA required for Axiom arrays?
All Axiom arrays (except for Axiom Genome-Wide Pan-African Array Set) require a total of 200 ng DNA. The Axiom Genome-Wide Pan-African Array Set requires a total of 300 ng DNA, or 100 ng DNA per array (there are 3 arrays).
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
What are the sample requirements for Axiom array processing at the Thermo Fisher Scientific Microarray Research Services Laboratory (MRSL)?
For information regarding our in-house MRSL requirements, please contact us via phone at 888-362-2447 (in North America) or via email at BioinformaticsServices@thermofisher.com.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.