Axiom™ Rice Genotyping Array
Applied Biosystems™

Axiom™ Rice Genotyping Array

Axiom Rice Genotyping Array offers comprehensive genomic coverage, which allows researchers to easily assay the natural variation of rice varietiesRead more
Have Questions?
Catalog NumberQuantity
5506251 plate
Catalog number 550625
Price (USD)
-
Quantity:
1 plate
Axiom Rice Genotyping Array offers comprehensive genomic coverage, which allows researchers to easily assay the natural variation of rice varieties worldwide to identify genes underlying important phenotypic traits (e.g., high yields), to understand genetic sources of variation, and to differentiate cultivars for fingerprinting and classification purposes.

Axiom Rice Genotyping Array includes 42,961 markers from GeneChip™ Rice 44K Genotyping Arrayand 5,499 markers from a whole-genome SNP array (RICE6K) for genomic breeding in rice.

Highlights:
• Content
   - Backwards compatibility with older data based on 44,000 makers from GeneChip Rice 44K SNP Genotyping Array
   - Detection of common genetic variants within and between the major subpopulations of rice, including indica, aus, tropical japonica, temperate japonica, and group V ('aromatic')
   - Efficient progeny screening in rice breeding with markers identified by Yu et al.

Applications:
• Complex-trait research
   - Flexible and fast identification and classification of germplasm
   - Identification of high-yield variants
   - Genome-wide association mapping and tracking of disease genes
• Molecular breeding
   - Allows for fingerprinting rice germplasm, genotyping bulked segregating pools, checking seed authenticity, and selecting genetic background
   - Enables genetic variation analysis of any O. sativa strain
   - Offers information for polymorphisms between indica and japonica subspecies as well as varieties within indica and japonica groups

Required Products
GeneChip Command Console Software
GeneTitan Multi-Channel Instrument
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)Microarray Analysis
Quantity1 plate
ArrayGenotyping
Format96-array Plate
Number of Arrays96 arrays
SpeciesRice
Unit Size1 plate

Frequently asked questions (FAQs)

What is the starting amount of DNA required for Axiom arrays?

All Axiom arrays (except for Axiom Genome-Wide Pan-African Array Set) require a total of 200 ng DNA. The Axiom Genome-Wide Pan-African Array Set requires a total of 300 ng DNA, or 100 ng DNA per array (there are 3 arrays).

Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.

What are the sample requirements for Axiom array processing at the Thermo Fisher Scientific Microarray Research Services Laboratory (MRSL)?

For information regarding our in-house MRSL requirements, please contact us via phone at 888-362-2447 (in North America) or via email at BioinformaticsServices@thermofisher.com.

Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.