Axiom™ Bovine-Ovine-Caprine Genotyping Array - FAQs
Axiom™ Bovine-Ovine-Caprine Genotyping Array - FAQs
View additional product information for Axiom™ Bovine-Ovine-Caprine Genotyping Array - FAQs (550627)
2 product FAQs found
What is the starting amount of DNA required for Axiom arrays?
All Axiom arrays (except for Axiom Genome-Wide Pan-African Array Set) require a total of 200 ng DNA. The Axiom Genome-Wide Pan-African Array Set requires a total of 300 ng DNA, or 100 ng DNA per array (there are 3 arrays).
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
What are the sample requirements for Axiom array processing at the Thermo Fisher Scientific Microarray Research Services Laboratory (MRSL)?
For information regarding our in-house MRSL requirements, please contact us via phone at 888-362-2447 (in North America) or via email at BioinformaticsServices@thermofisher.com.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.