Axiom™ Ovine Genotyping Array
Applied Biosystems™

Axiom™ Ovine Genotyping Array

The Applied Biosystems Axiom Ovine Genotyping Array provides the power and resolution to enable a wide range of applications inWeitere Informationen
Have Questions?
KatalognummerMenge
5508381 Platte
Katalognummer 550838
Preis (EUR)
-
Menge:
1 Platte
The Applied Biosystems Axiom Ovine Genotyping Array provides the power and resolution to enable a wide range of applications in sheep breeding, including breed assignment, parentage verification and discovery, and traceability and monitoring of polymorphisms of known large effect. The 11,000 markers on the array facilitate highly accurate imputation up to a higher density panel (e.g., 50K panel) The Axiom Ovine Genotyping Array includes 88 single nucleotide polymorphisms (SNPs) proposed by the International Sheep Genomics Consortium (ISGC) for parentage verification.

Features include:
• Expert design—developed in collaboration with Donagh Berry at Teagasc, Animal and Grassland Research and Innovation Center, Ireland
• Exceptionally high quality SNPs with excellent call and concordance rates across platforms and laboratories
• Includes SNPs for parentage as well as other traits including scrapie
• Compatible with Sheep Consortium 15K arrays and thus with previous studies

Applications
• Parentage verification and discovery
• Accurate genomic evaluations
• Genome-wide associations following accurate imputation to higher density panel
• Breed assignment
• Monitoring of major genes
• Aneuploidy detection—early detection of such abnormalities enables identification of animals that are not suitable parents for the next generation • Measurement of genetic diversity including calculation of genomic inbreeding statistics
• Mating designs
• Traceability

Required products
GeneChip Command Console Software
GeneTitan Multi-Channel Instrument
Axiom Analysis Suite Software

Nur für Forschungszwecke. Nicht zur Verwendung bei diagnostischen Verfahren.

Specifications
Zur Verwendung mit (Anwendung)Microarray-Analyse
ProduktlinieAxiom
Menge1 Platte
ArrayDNA
FormatArray Plate
Anzahl Arrays384 arrays
Unit Size1 plate
Inhalt und Lagerung
One plate containing 384 arrays

Häufig gestellte Fragen (FAQ)

What is the starting amount of DNA required for Axiom arrays?

All Axiom arrays (except for Axiom Genome-Wide Pan-African Array Set) require a total of 200 ng DNA. The Axiom Genome-Wide Pan-African Array Set requires a total of 300 ng DNA, or 100 ng DNA per array (there are 3 arrays).

Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.

What are the sample requirements for Axiom array processing at the Thermo Fisher Scientific Microarray Research Services Laboratory (MRSL)?

For information regarding our in-house MRSL requirements, please contact us via phone at 888-362-2447 (in North America) or via email at BioinformaticsServices@thermofisher.com.

Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.