Product Image
Thermo Scientific™

Matrix™ SerialMate™ Pipette Tips, 220 μL, Extended Length, Sterile

Perform pipetting more effectively with Thermo Scientific™ Matrix™ SerialMate™ Pipette Tips, for use with the Matrix SerialMate Pipetter.
Have Questions?
Catalog NumberVolume (Metric)
5562220 μL
Catalog number 5562
Price (USD)
-
Volume (Metric):
220 μL
Perform pipetting more effectively with Thermo Scientific™ Matrix™ SerialMate™ Pipette Tips, for use with the Matrix SerialMate Pipetter.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter.Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
DescriptionDeep-Well Tips
For Use With (Equipment)Thermo Scientific Matrix SerialMate
FormatRack
Product LineMatrix SerialMate
SterilitySterile
Volume (Metric)220 μL
Unit SizeEach