Hamilton™ Reagent Reservoirs, 60 mL
Thermo Scientific™
Hamilton™ Reagent Reservoirs, 60 mL
These reagent reservoirs are for use with the NIMBUS Presto System. The special bottom geometry minimizes residual volumes. They are詳細を見る
製品番号(カタログ番号)数量
56694-011 x 28 reservoirs
製品番号(カタログ番号) 56694-01
価格(JPY)
-
数量:
1 x 28 reservoirs
These reagent reservoirs are for use with the NIMBUS Presto System. The special bottom geometry minimizes residual volumes. They are made of polypropylene and manufactured under clean room conditions. They are certified CE and IVD-compliant.
仕様
パッケージIndividually wrapped
数量1 x 28 reservoirs
容量(メートル法)60 mL
材料Polypropylene
無菌性Non-sterile
タイプTrough
Unit SizeEach