What is the diameter of the membrane in Nalgene Rapid-Flow Sterile Disposable Filter Units with PES, CN, SFCA or Nylon Membranes?
The diameter of the membrane is 90 mm.
Search
View additional product information for Nalgene™ Rapid-Flow™ Sterile Disposable Filter Units with PES Membrane, 500mL, 0.2µm pore, 90mm membrane - FAQs (5690020)
1 product FAQs found