Magnesium stearate (vegetable based), Hyqual™, GenAR™, NF, GMP, Macron Fine Chemicals™
Avantor™

Magnesium stearate (vegetable based), Hyqual™, GenAR™, NF, GMP, Macron Fine Chemicals™

This product is GMP Manufactured and meets N.F, ChP, BP, EP, and JP Chemical Specifications.
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
2.5 kg
12 kg
Catalog number 5712-06
Price (USD)
-
Quantity:
2.5 kg
This product is GMP Manufactured and meets N.F, ChP, BP, EP, and JP Chemical Specifications.
Specifications
CAS557-04-0
Chemical Name or MaterialMagnesium stearate
ColorWhite
Melting Point130–140°C
Molecular FormulaC36H70MgO4
PackagingMall STAKMOR case
SynonymStearic acid Magnesium(II) salt, Stearic acid magnesium salt, Magnesium(II) Stearate
GradeMulti-Compendial/NF
Molecular Weight (g/mol)591.25
Physical FormSolid
Quantity2.5 kg
Unit SizeEach