Do you offer tips for cloning siRNA templates into pSilencer vectors?
Please review these tips (http://tools.thermofisher.com/content/sfs/manuals/pSil_tips.pdf) for cloning siRNA templates into pSilencer vectors.
Search
View additional product information for PSILENCER 4.1-CMVHYGRO COIL - FAQs (5777M)
1 product FAQs found