Multidrop™ DW Reagent Dispenser, 100-120 V 50/60 Hz - FAQs
Multidrop™ DW Reagent Dispenser, 100-120 V 50/60 Hz - FAQs
View additional product information for Multidrop™ DW Reagent Dispenser - FAQs (5840177)
1 product FAQs found
Do I need specific plate adapters for the Multidrop Combi or MD Combi nL?
No, the Multidrop Combi and Combi nL incorporate a fixed plate adapter and does not use any external plate adapters.