Multidrop™ Combi Reagent Dispenser, Multidrop Combi with SMART 2 option - FAQs
Multidrop™ Combi Reagent Dispenser, Multidrop Combi with SMART 2 option - FAQs
View additional product information for Multidrop™ Combi Reagent Dispenser - FAQs (5840320)
5 product FAQs found
When should I use the Versette Automated Liquid Handler vs. the Multidrop Combi Reagent Dispenser?
The Versette Automated Liquid Handler is the perfect solution for applications that require aspiration of liquid, plate transfers/reformatting, mixing, serial dilutions, and other common plate preparations performed using hand held pipettes in the lab.
The Multidrop Combi Reagent Dispenser should be used for fast bulk reagent dispensing and plate preparation.
Where can I see the Multidrop Combi internal software version number?
When the instrument is powered on, the version number only appears on the bottom of the screen during the startup animation.
Can the Multidrop Combi and Multidrop Combi nL priming vessel be autoclaved?
Yes, autoclaving conditions are as follows: 1 bar pressure at 121 degrees C for 20 minutes.
What is the material of the Multidrop Combi and Multidrop Combi nL priming vessel?
The Multidrop Combi and Multidrop Combi nL priming vessel is made of blue polypropylene (PP).
Do I need specific plate adapters for the Multidrop Combi or MD Combi nL?
No, the Multidrop Combi and Combi nL incorporate a fixed plate adapter and does not use any external plate adapters.