Multidrop Combi SMART+ Dispenser - FAQs
Multidrop Combi SMART+ Dispenser - FAQs
View additional product information for Multidrop Combi SMART+ Dispenser - FAQs (5840340)
2 product FAQs found
Are the Multidrop Combi reagent dispensers compatible with Momentum Workflow Scheduling Software?
Yes, the Multidrop Combi reagent dispensers are compatible with the Momentum Software and can be integrated into automated workflow platforms. If you are planning to set up an automated workflow using the Momentum Software, you will also need a microplate mover such as the Orbitor RS2 Microplate Mover, Spinnaker Microplate Robot, or Spinnaker 3 Microplate Mover.
Do Multidrop Combi Reagent Dispensers (Cat. Nos. 58403xx and 58404xx) come with a laptop?
No, Multidrop Combi Reagent Dispensers are not sold with a laptop. This would need to be purchased separately. You can also use your own laptop or PC, as long as it meets the minimum PC requirements outlined in the Thermo Scientific FILLit Software for Multidrop Combi User Manual (pg 15).