Do the Multidrop Pico 1 & Pico 8 Digital Dispensers come with a laptop?
No, Multidrop Pico 1 & Pico 8 Digital Dispensers are not sold with a laptop. This would need to be purchased separately. You can also use your own laptop or PC, as long as it meets the minimum PC requirements outlined in Chapter 3 of the Thermo Scientific Multidrop Pico 1 or Multidrop Pico 8 Operating Manuals: