Product Image
Invitrogen™
eBioscience™ Phalloidin eFluor™ 520
Phalloidin is a peptide toxin isolated from the Amanita phalloides mushroom. Phalloidin selectively binds F-actin and functions to stablilize actinRead more
Have Questions?
Catalog NumberQuantity
59-6559-05300 tests
Catalog number 59-6559-05
Price (USD)
504.00
Each
Quantity:
300 tests
Price (USD)
504.00
Each
Phalloidin is a peptide toxin isolated from the Amanita phalloides mushroom. Phalloidin selectively binds F-actin and functions to stablilize actin polymers by inhibiting the dissociation of actin monomers from the filament ends. Because F-actin polymers compose the cytoskeleton of almost all eukaryotic cells, phalloidin conjugates can be useful probes for visualizing the structure of live or fixed cells using microscopy. Phalloidin eFluor™ 520 can be used to label cells of multiple species. Its exitation and emission is similar to Alexa Fluor 488 and FITC.
Phalloidin eFluor™ 520 is supplied as a frozen liquid. To prepare the 1000X stock solution, thaw the vial, add 30 uL high-quality anhydrous DMSO, mix and briefly spin down. The resulting solution (phalloidin DMSO stock) should be aliquoted, protected from light and moisture, and stored at -20°C. To prepare a 1X working solution of phalloidin conjugate, add 1 uL of 1000X phalloidin DMSO stock solution to 1 mL of PBS containing 1% BSA.
Emit
535 nm
Excite
475 nm
Reported Application
Immunocytochemistry, Immunohistochemical Staining, Cell Labeling, Microscopy
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Dye TypeeFluor 520
Excitation Wavelength Range488/520
No. of Tests300 tests
Product LineeBioscience
Quantity300 tests
Label TypeeFluor
Product TypePhalloidin
Sub Cellular LocalizationCytoskeleton
Unit SizeEach
Frequently asked questions (FAQs)
Can I combine Click-iT or Click-iT Plus reactions with phalloidin conjugates used for actin staining?
We do not recommend using phalloidin conjugates for staining actin in combination with traditional Click-iT or Click-iT Plus reactions since phalloidin is extremely sensitive to the presence of copper.
For staining actin in combination with traditional Click-iT or Click-iT Plus reactions, we recommend using anti-α-actin antibodies for staining actin in the cytoskeleton. You can find a list of our actin antibodies here.
Another option would be to use the Click-iT Plus Alexa Fluor Picolyl Azide Toolkit (Cat. Nos. C10641, C10642, C10643). These Click-iT Plus toolkits provide Copper and Copper protectant separately which makes it easier to titrate the copper concentration to obtain optimal labeling with minimal copper-mediated damage. You may need to optimize the click reaction with the lowest possible concentration of copper and then perform the phalloidin staining.
Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.