Nalgene™ Rapid-Flow™ Sterile Disposable Bottle Top Filters with PES Membrane, 500mL, 0.2μm pore, 33mm neck - FAQs
Nalgene™ Rapid-Flow™ Sterile Disposable Bottle Top Filters with PES Membrane, 500mL, 0.2μm pore, 33mm neck - FAQs
View additional product information for Nalgene™ Rapid-Flow™ Sterile Disposable Bottle Top Filters with PES, CN, SFCA or Nylon Membranes - FAQs (596-3320, 295-3345, 295-4545, 292-3320, 596-4520, 292-4520, 291-4545, 291-3345, 296-3345, 291-3320, 291-4520, 296-4545, 597-3320, 597-4520, 290-3320, 290-4520, 290-3345, 595-3320, 595-4520, 290-4545)
1 product FAQs found
Can I remove the filter membranes from the Nalgene Rapid-Flow Sterile Disposable Bottle Top Filters with PES, CN, SFCA or Nylon Membranes?
No, the filter membranes for the bottle top filters are molded in and cannot be removed.