Lindberg/Blue M™ Tube Furnace Adapters/Sleeves
Thermo Scientific™

Lindberg/Blue M™ Tube Furnace Adapters/Sleeves

Use Thermo Scientific™ Lindberg/Blue M™ Tube Furnace Adapters/Sleeves with Lindberg/Blue M Tube Furnaces.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTypeFor Use With (Equipment)
59545Tube Furnace Adapter, 3 in.STF55346C Tube Furnace
59510Tube Furnace Adapter, 0.75 in.HTF55122 Tube Furnace
59511Tube Furnace Adapter, 1 in.HTF55122 Tube Furnace
59519Tube Furnace Blank AdapterHTF55122 Tube Furnace
59522Tube Furnace Adapter, 1.5 inHTF55322, HTF55342, HTF55347 Furnace
59523Tube Furnace Adapter, 2 in.HTF55322, HTF55342, HTF55347 Furnace
59524Tube Furnace Adapter, 2.5 in.HTF55322, HTF55342, HTF55347 Furnace
59525Tube Furnace Adapter, 3 in.HTF55322, HTF55342, HTF55347 Furnace
59529Tube Furnace Blank AdapterHTF55322, HTF55342, HTF55347 Furnace
59535Tube Furnace Adapter, 3 in.HTF55667 Tube Furnace
59536Tube Furnace Adapter, 4 in.HTF55667 Tube Furnace
59537Tube Furnace Adapter, 5 in.HTF55667 Tube Furnace
59541Tube Furnace Adapter, 1 in.STF55346C Tube Furnace
59549Tube Furnace Blank AdapterSTF55346C Tube Furnace
59555Tube Furnace Adapter, 3 in.STF55666C Tube Furnace
59556Tube Furnace Adapter, 4 in.STF55666C Tube Furnace
59557Tube Furnace Adapter, 5 in.STF55666C Tube Furnace
59558Tube Furnace Adapter, 6 in.STF55666C Tube Furnace
7219-2134-001Tube Furnace Sleeve, 1 in.STF54434C Tube Furnace
7219-2134-002Tube Furnace Sleeve, 2 in.STF54434C Tube Furnace
7219-2134-003Tube Furnace Sleeve, 3 in.STF54434C Tube Furnace
7219-2134-011Tube Furnace Sleeve, 3 in.STF54454C Tube Furnace
7219-2134-012Tube Furnace Sleeve, 2 in.STF54454C Tube Furnace
7219-2134-013Tube Furnace Sleeve, 1 in.STF54454C Tube Furnace
7219-2147-002Tube Furnace Adapter, 2 in.STF54434C Tube Furnace
7219-2147-003Tube Furnace Adapter, 3 in.STF54434C Tube Furnace
7219-2147-012Tube Vestibule, 2 in.STF54454C Tube Furnace
7219-2147-013Tube Vestibule, 1 in.STF54454C Tube Furnace
Catalog number 59545
Price (USD)
-
Type:
Tube Furnace Adapter, 3 in.
For Use With (Equipment):
STF55346C Tube Furnace
Use Thermo Scientific™ Lindberg/Blue M™ Tube Furnace Adapters/Sleeves with Lindberg/Blue M Tube Furnaces.
Specifications
Diameter (Metric) Process Tube76.2 mm
For Use With (Equipment)STF55346C Tube Furnace
TypeTube Furnace Adapter, 3 in.
Unit SizeEach