Nalgene™ Unwire™ Polypropylene Test Tube Racks, for 13mm tubes, red - FAQs
Nalgene™ Unwire™ Polypropylene Test Tube Racks, for 13mm tubes, red - FAQs
View additional product information for Nalgene™ Unwire™ Polypropylene Test Tube Racks - FAQs (5976-0316PK, 5976-0013PK, 5976-0513PK, 5976-0516PK, 5976-0313PK, 5976-0416, 5976-0016PK, 5976-0316, 5976-0513, 5976-0413, 5976-0516, 5976-0413PK, 5976-0016, 5976-0313, 5976-0416PK, 5976-0013)
2 product FAQs found
Which Unwire racks are autoclavable?
Any of our ResMer polyketone test tube racks can be autoclaved, by our standard validated parameters (121 degrees Celsius at 15 psig for 20 minutes). Due to their molded structure, our polypropylene Unwire racks (unlike our other polyproylene labware products) cannot be autoclaved.
Which of your Unwire test tube racks can be used for 15 mL or 50 mL sterile conical centrifuge tubes?
Any of our Unwire racks with spaces for tubes 20 mm and 30 mm in diameter can be used, respectively, for 15 mL and 50 mL sterile conical centrifuge tubes.