HyperSep™ Retain PEP Cartridges
Thermo Scientific™

HyperSep™ Retain PEP Cartridges

Get balanced retention for polar and nonpolar analytes with Thermo Scientific™ HyperSep™ Retain PEP Cartridges.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberBed WeightColumn Capacity (Metric)
60107-207100 mg6 mL
60107-20130 mg1 mL
60107-20360 mg3 mL
60107-204200 mg3 mL
60107-205500 mg3 mL
60107-206500 mg6 mL
60107-20860 mg6 mL
60107-211150 mg6 mL
60107-212200 mg6 mL
60107-2151000 mg25 mL
60107-217100 mg3 mL
60107-2181000 mg6 mL
Catalog number 60107-207
Price (USD)
287.82
Pack of 30
Bed Weight:
100 mg
Column Capacity (Metric):
6 mL
Price (USD)
287.82
Pack of 30
Get balanced retention for polar and nonpolar analytes with Thermo Scientific™ HyperSep™ Retain PEP Cartridges, packed with a high capacity, high purity, highly porous polystyrene DVB material modified with urea functional groups. With chemistry pH stable 0 to 14, the of these cartridges in SPE sample preparation can significantly enhance the robustness and reliability of many biological, pharmaceutical, forensic and environmental applications.
Features:
Ideal for large samples, Hypersep Retain cartridges conform to industry standard configurations and allow efficient routine processing by vacuum, positive pressure or centrifugation.
  • High, consistent recoveries
  • Fast and easy sample preparation and method development
  • Reproducible, reliable sample preparation in a traditional format
  • Available in a wide range of chemistries and bed weights
Applications:
  • Drugs and metabolites in biological fluids
  • Peptides in serum, plasma or biological fluids
  • Environmental samples
For Use With: The Thermo Scientific™ HyperSep-96™ System
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Column Capacity (Metric)6 mL
Description100 mg bed weight; 6 mL column volume
For Use With (Application)Retention of Polar and Nonpolar Analytes
QuantityPack of 30
Bed Weight100 mg
For Use With (Equipment)Thermo Scientific HyperSep-96 System
Particle Size40 to 60 μm
Product LineHyperSep
SorbentHyperSep Urea-Modified Polystyrene DVB
TypeSolid Phase Extraction Cartridge
Volume (Metric)6 mL
Unit SizePack of 30