SMART Digest™ Chymotrypsin Kits
Thermo Scientific™
SMART Digest™ Chymotrypsin Kits
Obtain fast, simple, and highly reproducible chymotrypsin protein digestion for peptide characterization and quantitation in biopharmaceutical research.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescription
60109-105-BBulk Resin option with Filter/Collection plate
60109-104with Collection plate
60109-104-MB-LPHLow pH, Magnetic Beads, Bulk option for MAM workflows
60109-104-BBulk Resin option
60109-104-MBMagnetic Bulk Resin option
60109-105with Filter/Collection plate
60109-105-MBMagnetic Bulk Resin option with Filter/Collection plate
60109-106SMART Digest Chymotrypsin Kit with SOLAμ/collection plate
60109-106-BBulk Resin option with SOLAμ/collection plate
60109-106-MBMagnetic Bulk Resin option with SOLAμ/collection plate
Catalog number 60109-105-B
Price (USD)
1,063.30
Each
Description:
Bulk Resin option with Filter/Collection plate
Price (USD)
1,063.30
Each
Obtain high-quality analytical results from protein digests using Thermo Scientific™ SMART Digest™ Chymotrypsin Kits. A significant advance in sample preparation for biopharmaceutical protein research, these kits provide fast and simple protein digestion with high reproducibility, high sensitivity, and high levels of data quality in a format that's compatible with automation. SMART Digest Kits are a significant improvement over in-solution protein digestion technologies, which are not reproducible, have poor sensitivity, and employ protracted methodologies not amenable to automation.
Chymotrypsin Protein Digestion
Characterize and quantitate proteins faster, easier, and quicker than ever before using SMART Digest kits. This version of the kit uses chymotrypsin, which cleaves peptide bonds formed by aromatic residues such as tyrosine, phenylalanine, and tryptophan.
Easy and Simple
  • Significantly increased speed of sample processing
  • Simple process
  • Easily automate entire protocol for high-throughput processing
Reliable Results
  • Increased reproducibility over existing protocols
  • Higher sensitivity
  • Higher data confidence
Significantly Reduces Sample Preparation Time
Typically it takes less than 60 minutes (dependent on sample complexity) to achieve full digestion, whereas an in-solution digestion can take up to 24 hours.
Cleaner Samples
Reduction in the number of chemicals used and the immobilized chymotrypsin design helps mitagate against chemically induced post translational modifications (PTMs) and autolysis. The results are easier to interpret.
Couple with SOLAμ SPE plates for desalting
When used with the SMART Digest for desalting, Thermo Scientific™ SOLAμ™ SPE solid phase extraction plates provide a fast, easy-to-use, robust, and easily automatable sample preparation solution for analysis of biomolecules.
Specifications
DescriptionBulk Resin option with Filter/Collection plate
EnzymeChymostrypsin
For Use With (Application)Peptide Quantitation, Peptide Mapping
IncludesBulk Non-magnetic SMART Digest Resin, SMART Digest Buffer, Collection Plate, Filter Plate
CaptureNone
CleavagePeptide bonds of aromatic residues (e.g., tyrosine, phenylalanine, tryptophan)
FormatSMART Digest Resin, Bulk Non-magnetic
Product LineSMART Digest
TypeProtein Digestion
Unit SizeEach