SMART Digest™ Pepsin Kits
Thermo Scientific™
SMART Digest™ Pepsin Kits
Use these pepsin protein digestion kits in your peptide mapping and peptide quantitation workflows to obtain high quality analytical results.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescription
60109-111with Filter and Collection Plate
60109-110with Collection Plate
60109-112with SOLAμ Collection Plate
60109-110-MBwith Magnetic Bead, Bulk Option (No Collection Plate)
Catalog number 60109-111
Price (USD)
1,047.48
Each
Description:
with Filter and Collection Plate
Price (USD)
1,047.48
Each
Use Thermo Scientific™ SMART Digest™ Pepsin Kits in your peptide mapping and peptide quantitation workflows to obtain high quality analytical results. A significant advance in sample preparation for biopharmaceutical protein research, the kits provide fast and simple protein digestion with high reproducibility, high sensitivity, and high levels of data quality in a format that's compatible with automation. The SMART Digest Pepsin Kits are a significant improvement over current in-solution protein digestion technologies, which are not reproducible, have poor sensitivity, and employ protracted methodologies not amenable to automation.
How SMART Digest Pepsin Kits work
Pepsin cleaves proteins preferentially at carboxylic groups of aromatic amino acids such as phenylalanine, tryptophan and tyrosine. It will not cleave at bonds containing valine, alanine or glycine. Pepsin is often used to generate antibody fragments. Pepsin is used to generate F(ab')2 fragments by removing only that portion of the Fc domain beyond the hinge region. The resulting F(ab')2 fragments are composed of two antibody-binding Fab' fragments connected by disulfides in the hinge region.
Specifications
Descriptionwith Filter and Collection Plate
EnzymePepsin
For Use With (Application)Peptide Quantitation, Peptide Mapping
IncludesSMART Digest Resin Pre-filled into 96 PCR Tubes, Collection Plate, Filter Plate
CaptureNone
CleavageHydrophobic residues in P1 and P1' positions at the carboxyl side of phenylalanine and leucine and at the carboxyl side of glutamic acid
FormatPre-filled PCR Tubes
Product LineSMART Digest
TypeDigest Pepsin Kit
Unit SizeEach