SMART Digest™ Bulk Fractionation Kits: Protein A
Thermo Scientific™
SMART Digest™ Bulk Fractionation Kits: Protein A
Use this kit to for purification of immunoglobulins prior to protein A digestion.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescription
60114-104Magnetic, Soluble
60114-101Non-Magnetic, Soluble
60114-102Non-Magnetic, with SOLAμ Collection Plate, Soluble
60114-103Magnetic, with SOLAμ Collection Plate, Soluble
60116-101Magnetic, Standalone
Catalog number 60114-104
Price (USD)
1,704.48
Each
Description:
Magnetic, Soluble
Price (USD)
1,704.48
Each
Obtain fast, simple, highly reproducible, and sensitive immunocapture using the high loading capacity of the Thermo Scientific™ SMART Digest™ Bulk Fractionation Kits: Protein A. These kits were designed to remove the challenges often associated with the process of protein clean up by providing a simple and easy capture workflow.
The SMART Digest Bulk fractionation Protein A Kits contain beads (either non-magnetic or magnetic, the magnetic allowing automation) which target purification of immunoglobulins due to the specific and strong affinity for binding to the Fc portion of the molecule. The unwanted interferences are then removed leaving a clean sample containing the target molecule. Once completed subsequent digestion can be facilitated with SMART Digest enzymes if required.
Specifications
DescriptionMagnetic, Soluble
EnzymeTrypsin
For Use With (Application)Immunocapture and Protein Digest Sample Preparation
IncludesBulk Magnetic SMART Digest Soluble Resin
CaptureProtein A
CleavageC-terminal side of lysine and arginine amino acids
FormatBulk Magnetic
Product LineSMART Digest
TypeBulk Fractionation Kit
Unit SizeEach