SMART Digest™ Bulk Fractionation Kits: Protein G
Thermo Scientific™
SMART Digest™ Bulk Fractionation Kits: Protein G
Obtain fast, simple, highly reproducible, and sensitive immunocapture using the high loading capacity of the Thermo Scientific SMART Digest Bulk Fractionation Kits: Protein G.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescription
60115-101Non-Magnetic, Soluble
60115-102Non-Magnetic, with SOLAμ Collection Plate, Soluble
60115-103Magnetic, with SOLAμ Collection Plate, Soluble
60115-104Magnetic, Soluble
60117-101Magnetic, Standalone
Catalog number 60115-101
Price (USD)
1,487.53
Each
Description:
Non-Magnetic, Soluble
Price (USD)
1,487.53
Each
Obtain fast, simple, highly reproducible, and sensitive immunocapture using the high loading capacity of the Thermo Scientific™ SMART Digest™ Bulk Fractionation Kits: Protein G. These kits remove the challenges often associated with protein cleanup by providing a simple and easy capture workflow.
SMART Digest Bulk Fractionation Kits: Protein G contain beads that target purification of immunoglobulins due to the specific and strong affinity for binding to the Fab and Fc portions of the molecule. Choose from non-magnetic beads, or magnetic beads which enable automation. The unwanted interferences are then removed leaving a clean sample containing the target molecule. Once complete, perform digestion using SMART Digest enzymes if required.
Specifications
DescriptionNon-Magnetic, Soluble
EnzymeTrypsin
For Use With (Application)Immunocapture and Protein Digest Sample Preparation
IncludesBulk Non-magnetic SMART Digest Soluble Resin
CaptureProtein G
CleavageC-terminal side of lysine and arginine amino acids
FormatBulk Non-magnetic
Product LineSMART Digest
TypeSMART Digest Bulk Fractionation Kit
Unit SizeEach