SMART Digest™ Bulk Fractionation Kits: Protein A
Thermo Scientific™
SMART Digest™ Bulk Fractionation Kits: Protein A
Use this kit to for purification of immunoglobulins prior to protein A digestion.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescription
60116-101Magnetic, Standalone
60114-101Non-Magnetic, Soluble
60114-102Non-Magnetic, with SOLAμ Collection Plate, Soluble
60114-103Magnetic, with SOLAμ Collection Plate, Soluble
60114-104Magnetic, Soluble
Catalog number 60116-101
Price (USD)
864.00
Each
Description:
Magnetic, Standalone
Price (USD)
864.00
Each
Obtain fast, simple, highly reproducible, and sensitive immunocapture using the high loading capacity of the Thermo Scientific™ SMART Digest™ Bulk Fractionation Kits: Protein A. These kits were designed to remove the challenges often associated with the process of protein clean up by providing a simple and easy capture workflow.
The SMART Digest Bulk fractionation Protein A Kits contain beads (either non-magnetic or magnetic, the magnetic allowing automation) which target purification of immunoglobulins due to the specific and strong affinity for binding to the Fc portion of the molecule. The unwanted interferences are then removed leaving a clean sample containing the target molecule. Once completed subsequent digestion can be facilitated with SMART Digest enzymes if required.
Specifications
DescriptionMagnetic, Standalone
EnzymeNone
For Use With (Application)Immunocapture and Sample Preparation
IncludesBulk Magnetic SMART Digest Resin
CaptureProtein A
FormatBulk Magnetic
Product LineSMART Digest
TypeBulk Fractionation Kit
Unit SizeEach