SMART Digest™ Bulk Fractionation Kits: Protein G
Thermo Scientific™
SMART Digest™ Bulk Fractionation Kits: Protein G
Obtain fast, simple, highly reproducible, and sensitive immunocapture using the high loading capacity of the Thermo Scientific SMART Digest Bulk Fractionation Kits: Protein G.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescription
60117-101Magnetic, Standalone
60115-101Non-Magnetic, Soluble
60115-102Non-Magnetic, with SOLAμ Collection Plate, Soluble
60115-103Magnetic, with SOLAμ Collection Plate, Soluble
60115-104Magnetic, Soluble
Catalog number 60117-101
Price (USD)
864.00
Each
Description:
Magnetic, Standalone
Price (USD)
864.00
Each
Obtain fast, simple, highly reproducible, and sensitive immunocapture using the high loading capacity of the Thermo Scientific™ SMART Digest™ Bulk Fractionation Kits: Protein G. These kits remove the challenges often associated with protein cleanup by providing a simple and easy capture workflow.
SMART Digest Bulk Fractionation Kits: Protein G contain beads that target purification of immunoglobulins due to the specific and strong affinity for binding to the Fab and Fc portions of the molecule. Choose from non-magnetic beads, or magnetic beads which enable automation. The unwanted interferences are then removed leaving a clean sample containing the target molecule. Once complete, perform digestion using SMART Digest enzymes if required.
Specifications
DescriptionMagnetic, Standalone
EnzymeNone
For Use With (Application)Immunocapture and Sample Preparation
IncludesBulk Magnetic SMART Digest Resin
CaptureProtein G
FormatBulk Magnetic
Product LineSMART Digest
TypeSMART Digest Bulk Fractionation Kit
Unit SizeEach