SMART Digest™ RNase Kits
Thermo Scientific™
SMART Digest™ RNase Kits
Use SMART Digest™ RNase Kits in your oligonucleotide mapping and quantitation workflows to obtain high quality analytical results.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberEnzyme
60120-102RNase A
60120-101RNase T1
Catalog number 60120-102
Price (USD)
1,443.55
Each
Enzyme:
RNase A
Price (USD)
1,443.55
Each
Obtain high-quality analytical results from mRNA digestions using Thermo Scientific™ SMART Digest™ RNase Kits. A significant advance in sample preparation for oligonucleotide mapping and quantitation, these kits provide fast and simple mRNA digestion with high reproducibility, high sensitivity, and high levels of data quality in a format that is compatible with automation.
These RNase kits use optimized, heat stable enzymes on magnetic beads, allowing for control of partial digestion to produce required fragments of ideal length for sequencing by HRAM-MS.
Easy and Simple
  • A simple process and control of digestion time ensure required fragment length for comprehensive mRNA sequencing by HRAM-MS
  • Easily automate the entire protocol for high-throughput processing
  • Single nuclease digestion allows full characterization of mRNA
Reliable Results
  • Increased reproducibility over existing protocols
  • Higher data confidence
  • No nuclease contamination of the analytical column
RNase kits are available with with two endoribonuclease enzyme specificities:
  • RNase T1 degrades single-stranded RNA at G residues
  • RNase A degrades single-stranded RNA at C and U residues
Specifications
CleavageDegrades single-stranded RNA at C and U residues
For Use With (Application)Oligonucleotide Mapping
FormatBulk Magnetic SMART Digest Resin
IncludesBulk Magnetic SMART Digest Resin, SMART Digest Buffer
Product LineSMART Digest
TypeRNA Digestion
EnzymeRNase A
Unit SizeEach