Electronic Vial Crimpers and Decrimpers
Thermo Scientific™
Electronic Vial Crimpers and Decrimpers
Attach or remove crimp vial seals with high sample security and a simple, one-handed push of a button by using these Electronic Vial Crimpers and Decrimpers.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSize
60180-ECRH11KIT11 mm
60180-ECRH13KIT13 mm
60180-ECRH20KIT20 mm
Catalog number 60180-ECRH11KIT
Price (USD)
3,037.50
Each
Size:
11 mm
Price (USD)
3,037.50
Each
Attach or remove crimp vial seals with high sample security with a simple, one-handed push of a button. Thermo Scientific™ Electronic Vial Crimpers and Decrimpers are available as handheld and high-power models for 8mm, 11mm, 13mm, and 20mm crimp vial caps.
The electronic crimper's message window provides all-important information at one glance, such as:
  • The actual crimp power
  • An error message after an incorrect crimping process
  • Statistics and log of recent crimping results
  • In the language of your choice (English, Chinese, Japanese, German, French, Spanish, Italian, Korean)
These Electronic Vial Crimpers also feature adjustable settings for best crimp results and a motor with extended lifetime and no particle emission.
Electronic High-Power Crimpers
  • Recommended for steel caps because it provides highest crimping force independent from battery charge status
  • Includes a 12-volt DC supply with power cord
  • Kits include Crimp and Decrimp Heads; they are also available separately
  • Can be used as a hand-held device
  • Optional adjustable accessory base for added convenience
Electronic Hand-Held Crimpers and Decrimpers
  • For crimping of vials with aluminum, pressure-release, and bi-metal caps
  • The battery charge icon on the color screen changes to red when battery is low
  • Includes a fully rechargeable lithium ion battery and a universal power supply/recharger with international plug adapters
Maximized sample security with electronic crimping tools
  • Vial crimping and decrimping can be performed in most sample trays.
  • The crimp head selection also includes special crimp heads for 13mm and 20mm flip-off caps; more specials are available on request.
Specifications
Description11 mm Electronic High Power Crimper Kit
For Use With (Equipment)11 mm aluminum, bi-metal, and steel caps
IncludesElectronic High Power Tool, 12 volt DC power supply with power cord, 11 mm Crimp Head, 11 mm Decrimp Head
Compatible Storage TubesTubes with 11 mm Crimp Caps
Decapping FormatElectronic - Manual/Programmable
OperationProgrammable or Manual Hand-Held Operation
Size11 mm
Unit SizeEach