SureSTART™ WebSeal™ 96-Well Microtiter Plates, Round V-Bottom, Level 1 Everyday Analysis
Thermo Scientific™

SureSTART™ WebSeal™ 96-Well Microtiter Plates, Round V-Bottom, Level 1 Everyday Analysis

Use SureSTART™ WebSeal™ 96-Well Microtiter Plates, Round V-Bottom, Performance Level 1, for everyday chromatography analysis. They are Specification Certified and conform to ANSI-SLAS 4-2004 standards.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTotal Volume (Metric) WellNo. per PackVolume (Metric) WorkingHeight (Metric)
60180-P132500 μL120 Microplates20 to 450 μL14.5 mm
60180-P100450 μL20 Plates10 to 400 μL14.4 mm
60180-P130450 μL120 Plates10 to 400 μL14.4 mm
60180-P102500 μL10 Microplates20 to 450 μL14.5 mm
Catalog number 60180-P132
Price (USD)
506.01
Pack of 120
Total Volume (Metric) Well:
500 μL
No. per Pack:
120 Microplates
Volume (Metric) Working:
20 to 450 μL
Height (Metric):
14.5 mm
Price (USD)
506.01
Pack of 120

Thermo Scientific™ SureSTART™ Level 1 WebSeal™ 96-Well Round V-Bottom Microtiter Plates are designed for everyday chromatography analysis.

They provide an affordable choice for use with most HPLC/UHPLC instrument types and are ideal for non-regulated laboratories using water and polar solvents.

These 96-well plates are specification certified to ensure autosampler compatible physical dimensions and conform to ANSI-SLAS 4-2004 standards. Seal these well plates with SureSTART WebSeal 96-Well Plate Sealing Mats, 8 mm Diameter, Round, Level 1.

Performance Level 1 Plates – for Everyday Analysis

  • Ideal for non-regulated labs that use water and polar solvents
  • Commodity plates, seals, tapes
  • GC tested
  • Suitable for HPLC/UHPLC applications
  • Suitable for biopharmaceutical applications
  • Suitable for QA/QC applications using core detectors
  • Ideal for verifying compound presence and quantification using detection techniques such as UV, DAD/PDA, RI, Fluorescence, and ELSD
  • All Thermo Scientific SureSTART WebSeal well plates are dimensionally verified

Lot Traceable

  • 100% traceable through the manufacturing process
  • Certificate of Conformance available by lot number upon request

About Thermo Scientific WebSeal Well Plates & Mats

Thermo Scientific WebSeal plates and mats are available in three performance levels:

  • Level 1 well plates are for everyday applications and are suitable for non-regulated labs using water and polar solvents
  • Level 2 well plates are for high-throughput applications and are suitable for regulated labs using water and polar solvents
  • Level 3 well plates are for high performance applications and are suitable for regulated labs using all solvent types including non-polar solvents
Specifications
CertifiedSpecification Certified, GC/MS Tested
Diameter (Metric) Well8 mm
Height (Metric)14.5 mm
No. per Pack120 Microplates
Plate SizeMicrotiter
Product TypeMicroplate
Temperature Range (Metric)-80°C to 121°C
Total Volume (Metric) Well500 μL
Volume (Metric) Working20 to 450 μL
Array8 x 12
BarcodeNo Barcode
ColorClear
MaterialPolypropylene
No. of Wells96
Product LineSureSTART WebSeal Level 1
SterilityNon-sterile
Surface TreatmentNon-coated
Well ShapeRound, Conical Bottom (V-well)
Unit SizePack of 120

Frequently asked questions (FAQs)

Are bar-coded WebSeal plates available?

Yes, bar-coded WebSeal plates are available as a catalog product for the famly of Thermo Scientific Vanquish UHPLC machines only. However, plates can be provided with a pre-printed adhesive label applied. Contact your sales representative to assist with completing a form where you can define the bar code information and location of application onto the plate. Additional information required includes the physical requirements for the label such as temperature stability, solvent stability, etc.

What is the best way to seal a WebSeal Plate plus or plates with glass or PTFE inserts that need to be incubated at elevated temperatures?

WebSeal Plate+ or plates with glass or PTFE inserts should never be sealed with tapes or adhesive foils. Silicone mats should be used with the WebSeal Plate&plus products, preferably with a protective PTFE film to give increased chemical protection.

The glass inserted plates are sealed with specific silicone plug mats or individual caps. Again, most have additional PTFE films of layers to give increased chemical protection.

These plates, inserts, and mats were originally designed for combinatorial chemistry techniques with in situ chemical derivatization. Use with elevated temperatures is possible with the inserts and plugs. These are the closest plate format equivalent to glass autosampler vial products.

How do the Thermo Scientific-WebSeal Plate plus products compare to standard and certified products?

The WebSeal Plate+ coating provides additional surface protection that further reduces the possibility of interactions with solvents, sample component, or other materials. WebSeal Plate+ microptates display ultra-low background extractable organics profiles that compare favorably to our mass spectrometry certified vial products.

How do WebSeal-Certified low bleed plates compare to others?

Well plates are manufactured in molds. In order to ''force'' the ready produced plate to come out of this mold for the next shot, some manufacturers use release agents (organic compounds that act as lubricants). These release agents can be detected in your chromatogram. The grade of polypropylene used by most plate manufacturers can contain monomers or other added ingredients that are susceptible to leaching on contact with organic solvents. WebSeal-Certified products are manufactured from high purity resins and use a process that does not require mold release agents and that is why they exhbit no significant background extractables.

Do standard WebSeal plates bleed with organic solvents, or is it better to purchase certified plates?

If you are using high concentrations of organic solvents in your sample, then you may be concerned about ''bleed''. This is a term which normally implies a constant loss of material from the column or separation system which leads to high background levels, increased noise, and loss of sensitivity. If using solutions with a high level of buffer or water, then this should not be a concern as such components will not leach under these conditions. All plates may be used.

The evaluation of the plastic resins used in the production of plates was also carried out by controlled extraction of leachable components with organic solvents associated with HPLC and GC operation. These included methanol, acetonitrile, and hexane. Standard plates showed low level of extractables when tested at the highest instrument sensitivity range. These extractables found are characteristic of the common materials used for microplates in the clinical market and plates that have been used for general purpose chromatography applications. This indicates that for many non-critical applications and with higher concentrations of sample, the standard quality plates will be acceptable. These plates are termed Chromatography Tested.

The very low levels of background extractables found during our testing of some plates showed for applications run at more sensitive instrument settings. Where higher purity of the plate is required, you should directly switch to the Certified plates as they are the lowest extraction PP plates currently available on the world market. For critical analysis with high organic solvent concentrations and low sample concentrations, the Certified product range should be the plates of choice.