EasyStart™ PCR Mix-in-a-Tube
Thermo Scientific™

EasyStart™ PCR Mix-in-a-Tube

Reduce tedious PCR set-up protocols to a single step using Thermo Scientific™ EasyStart™ PCR Mix-in-a-tube.
Have Questions?
Catalog NumberProduct Type
6028TS
also known as 6028
Tube
Catalog number 6028TS
also known as 6028
Price (USD)
-
Product Type:
Tube
Reduce tedious PCR set-up protocols to a single step using Thermo Scientific™ EasyStart™ PCR Mix-in-a-tube.

Features:

  • Delivers better specificity and yield than conventionally prepared mixtures
  • Compatible with a wide range of enzymes
  • Wax layer provides safe, easy storage of reagent mixture, prevents oxidation and evaporation, and eliminates wasted reagents

Ordering Information:

Use 0.2mL tubes for 0.2mL heating blocks and 0.5mL tubes for 0.5 or 0.6mL heating blocks.

Specifications
Length (Metric)19 cm
Product LineEasyStart Micro 20
Reaction Volume15 to 20 μL
Storage RequirementsTubes can be stored at ambient temperatures for extended periods; To remove PCR product, simply pierce the wax layer with a pipet tip
Product TypeTube
Volume (Metric)0.2 mL
Unit SizeEach