HyperSep™ C8 Plates
Thermo Scientific™
HyperSep™ C8 Plates
Need a less retentive alternative to C18 for polar and nonpolar compounds? These hydrophobic reversed phase C8 well plates offer excellent performance.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberBed WeightVolume (Metric) Well
60300-44625 mg1 mL
60307-21110 mg2 mL
60307-21225 mg2 mL
60307-21350 mg2 mL
60300-44510 mg1 mL
60300-44750 mg1 mL
60300-448100 mg1 mL
Catalog number 60300-446
Price (USD)
519.36
Each
Bed Weight:
25 mg
Volume (Metric) Well:
1 mL
Price (USD)
519.36
Each
The use of SPE sample preparation can significantly enhance the robustness and reliability of many biological, pharmaceutical, forensic and environmental applications.
Thermo Scientific™ HyperSep™ C8 Plates offer a less retentive alternative to C18 for polar and nonpolar compounds, and are well-suited for methods requiring less retention than C18. The cartridges are ideally suited for the analysis of:
  • Drugs and their metabolites in biological samples
  • Peptides in biological samples
HyperSep C8 SPE Well Plates and Individual Wells provide excellent performance characteristics for moderately polar analytes. They also offer the potential for polar interactions greater than with C18.
HyperSep SPE Cartridges and Plates
HyperSep solid phase extraction cartridges offer reproducible, reliable sample preparation in a traditional format. Available in a wide range of chemistries and bed weights, the cartridges are ideal for large samples and conform to industry standard configurations, allowing efficient routine processing by vacuum, by positive pressure or centrifugation.
The Thermo Scientific™ HyperSep-96™ system incorporates traditional solid phase extraction sorbents into 96-well plate format providing fast analysis without sacrificing accuracy or reproducibility. The innovative design of HyperSep-96 well plates provides flexibility for both method development and scale adjustment to 96-well without method revalidation. Well plates are customizable, allowing any arrangement of wells with differing sorbent chemistries for simultaneous processing. HyperSep-96 products are either available in packs of 100 individual wells or in pre-assembled plates.
Specifications
Description96-Well Plate; Bed weight 25 mg
For Use With (Application)Screening of Drugs, Metabolites, and Peptides in Biological Samples
Bed Weight25 mg
Particle Size40 to 60 μm
Product LineHyperSep
SorbentHyperSep C8
TypeSolid Phase Extraction Well Plate
Volume (Metric) Well1 mL
Unit SizeEach