HyperSep™ Cyano Plates
Thermo Scientific™
HyperSep™ Cyano Plates
Get optimized isolation of polar compounds from nonpolar matrices, with these medium polarity cyano SPE well plates and individual wells.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberBed WeightVolume (Metric) Well
60300-82225 mg1 mL
60300-82110 mg1 mL
60300-82350 mg1 mL
60300-824100 mg1 mL
60307-304100 mg2 mL
Catalog number 60300-822
Price (USD)
528.28
Each
Bed Weight:
25 mg
Volume (Metric) Well:
1 mL
Price (USD)
528.28
Each
The use of SPE sample preparation can significantly enhance the robustness and reliability of many biological, pharmaceutical, forensic and environmental applications.
A medium polarity sorbent, Thermo Scientific™ HyperSep™ Cyano Plates are less retentive than silica or diol, and are optimized for the isolation of polar compounds from nonpolar matrices. Typical applications include:
  • Retention of polar compounds from hexane and oils
  • Reversed phase extraction of moderately polar compounds
HyperSep SPE Cartridges and Plates
HyperSep solid phase extraction cartridges offer reproducible, reliable sample preparation in a traditional format. Available in a wide range of chemistries and bed weights, the cartridges are ideal for large samples and conform to industry standard configurations, allowing efficient routine processing by vacuum, by positive pressure or centrifugation.
The HyperSep-96 system incorporates traditional solid phase extraction sorbents into 96-well plate format providing fast analysis without sacrificing accuracy or reproducibility. The innovative design of HyperSep-96 well plates provides flexibility for both method development and scale adjustment to 96-well without method revalidation. Well plates are customizable, allowing any arrangement of wells with differing sorbent chemistries for simultaneous processing. HyperSep-96 products are available in packs of 100 individual wells and in pre-assembled plates.
Specifications
DescriptionCyano 25 mg/1 mL 96 Removable Well Plate
For Use With (Application)Retention of Polar Compounds From Nonpolar Matrices
Quantity1 Pk.
Bed Weight25 mg
Particle Size40 to 60 μm
Product LineHyperSep
SorbentHyperSep Cyano
TypeSolid Phase Extraction Well Plate
Volume (Metric) Well1 mL
Unit SizeEach