HyperSep™ C18 Plates
Thermo Scientific™
HyperSep™ C18 Plates
Get improved retention for most nonpolar compounds and organic analytes in aqueous matrices with these hydrophobic reversed phase C18 well plates and individual wells.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescriptionBed Weight
60307-20296 Fixed Well Plate25 mg
60300-42596-Well Plate10 mg
60300-42696-Well Plate25 mg
60300-42796-Well Plate50 mg
60300-42896-Well Plate100 mg
60307-20496 Fixed Well Plate100 mg
60307-20396 Fixed Well Plate50 mg
60307-20196 Fixed Well Plate10 mg
Catalog number 60307-202
Price (USD)
528.28
Each
Description:
96 Fixed Well Plate
Bed Weight:
25 mg
Price (USD)
528.28
Each
The use of SPE sample preparation can significantly enhance the robustness and reliability of many biological, pharmaceutical, forensic and environmental applications. Thermo Scientific™ HyperSep™ C18 Plates feature a highly retentive alkyl-bonded phase for nonpolar to moderately polar compounds. The hydrophobic reversed phase material is retentive for most nonpolar compounds, and retains most organic analytes from aqueous matrices.
HyperSep SPE Cartridges and Plates
Thermo Scientific™ HyperSep™ solid phase extraction cartridges offer reproducible, reliable sample preparation in a traditional format. Available in a wide range of chemistries and bed weights, the cartridges are ideal for large samples and conform to industry standard configurations, allowing efficient routine processing by vacuum, by positive pressure or centrifugation.
The Thermo Scientific™ HyperSep-96™ system incorporates traditional solid phase extraction sorbents into 96-well plate format providing fast analysis without sacrificing accuracy or reproducibility. The innovative design of HyperSep-96 well plates provides flexibility for both method development and scale adjustment to 96-well without method revalidation. Well plates are customizable, allowing any arrangement of wells with differing sorbent chemistries for simultaneous processing. HyperSep-96 products are either available in packs of 100 individual wells or in pre-assembled plates.
Specifications
Description96 Fixed Well Plate
For Use With (Application)Extraction of Nonpolar to Moderately Polar Compounds
Quantity1 Pack
Volume (Metric) Well1 mL
Bed Weight25 mg
Particle Size40 to 60 μm
Product LineHyperSep
SorbentHyperSep C18
TypeSolid Phase Extraction Well Plate
Volume (Metric)1 mL
Unit SizeEach