HyperSep™ SCX Plates
Thermo Scientific™
HyperSep™ SCX Plates
Obtain excellent performance for extraction of charged basic compounds with these strong cation exchanger SPE well plates and individual wells.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescriptionBed Weight
60307-25396 Fixed Well Plate50 mg
60300-58896 Removable Well Plate100 mg
60307-25296 Fixed Well Plate25 mg
Catalog number 60307-253
Price (USD)
539.44
Each
Description:
96 Fixed Well Plate
Bed Weight:
50 mg
Price (USD)
539.44
Each
The use of SPE sample preparation can significantly enhance the robustness and reliability of many biological, pharmaceutical, forensic and environmental applications.
Thermo Scientific™ HyperSep™ SCX Plates feature a strong cation exchange sorbent for extraction of charged basic compounds. Typical applications include extraction of:
  • Antibiotics
  • Drug compounds
  • Organic bases
  • Amino acids
  • Catecholamines
  • Herbicides
HyperSep SCX Strong Cation Exchanger 96-Well Plates and Individual Wells are ideally suited for the extraction of a wide range of charged basic compounds.
  • Modified benzene ring provides potential for nonpolar interactions
  • Low pKa value
HyperSep SPE Cartridges and Plates
Thermo Scientific™ HyperSep™ solid phase extraction cartridges offer reproducible, reliable sample preparation in a traditional format. Available in a wide range of chemistries and bed weights, the cartridges are ideal for large samples and conform to industry standard configurations, allowing efficient routine processing by vacuum, by positive pressure or centrifugation.
The Thermo Scientific™ HyperSep-96™ system incorporates traditional solid phase extraction sorbents into 96-well plate format providing fast analysis without sacrificing accuracy or reproducibility. The innovative design of HyperSep-96 well plates provides flexibility for both method development and scale adjustment to 96-well without method revalidation. Well plates are customizable, allowing any arrangement of wells with differing sorbent chemistries for simultaneous processing. HyperSep-96 products are either available in packs of 100 individual wells or in pre-assembled plates.
Specifications
Description96 Fixed Well Plate
For Use With (Application)Extraction of Charged Basic Compounds
Quantity1 Pk.
Volume (Metric) Well1 mL
Bed Weight50 mg
Particle Size40 to 60 μm
Product LineHyperSep
SorbentHyperSep SCX
Type96-well Plate
Volume (Metric)2 mL
Unit SizeEach