Viper™ TQ Capillary Kits
Thermo Scientific™
Viper™ TQ Capillary Kits
Boost your productivity and confidence, and accelerate method development using application-specific LC workflows.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTypeMaterial
6036.2303BCapillary KitMP35N
6036.2010AFor Vanquish Inverse Gradient SystemsMP35N
6036.2020AFor Vanquish Tandem LC SystemsMP35N
6036.2030AFor Vanquish Trap/Simple Switch 2DLC SystemsMP35N
6036.2807AFor Vanquish Automatic Method Scouting SystemsMP35N
6036.2302ACapillary KitStainless Steel
Catalog number 6036.2303B
Price (USD)
-
Type:
Capillary Kit
Material:
MP35N
Improve chromatographic resolution by minimizing dead volume with the Thermo Scientific™ Viper™ TQ UHPLC Fittings. These pre-assembled kits allow you to quickly connect LC modules, valves, and columns for leak-free fluidic HPLC and UHPLC connections. Ideally suited for running application-specific workflows with your HPLC and LC-MS systems, like the Thermo Scientific™ Vanquish™ Method Development LC Systems, Thermo Scientific Vanquish Online 2D-LC Systems, or Thermo Scientific Vanquish Duo UHPLC Systems.
Enjoy the confidence of robust and reliable fingertight LC connections giving you tactile and audible feedback. Tailored to your advanced workflow requirements, Viper TQ application-specific capillary kits offer:
  • Simplified fluidic connection for all user levels with torque-controlled fingertight connections with sensory feedback
  • Easy-to-install, tool-free reusable capillaries requiring up to 40% less force compared to non-torque controlled fittings
  • Reliable, reproducible LC connections that deliver low carryover and simplify method transfer
Specifications
MaterialMP35N
Product LineViper TQ
TypeCapillary Kit
Unit SizeEach