Viper™ Automated Method Scouting Solution Kits
Actual product may vary
Viper™ Automated Method Scouting Solution Kits
Viper™ Automated Method Scouting Solution Kits
Viper™ Automated Method Scouting Solution Kits
Viper™ Automated Method Scouting Solution Kits
Viper™ Automated Method Scouting Solution Kits
Thermo Scientific™
Viper™ Automated Method Scouting Solution Kits
Obtain all the easy, zero-dead-volume fittings and capillaries needed for your method scouting LC system in this single kit.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Equipment)Diameter (Metric) Outer
6042.2807Dionex UltiMate 3000 BioRS System100 μm
6040.2807Dionex UltiMate 3000 Quaternary RS System130 μm
6040.2808Dionex UltiMate 3000 Quaternary SD System180 μm
6042.2808Dionex UltiMate 3000 BioRS System180 μm
Catalog number 6042.2807
Price (USD)
-
For Use With (Equipment):
Dionex UltiMate 3000 BioRS System
Diameter (Metric) Outer:
100 μm
Avoid data-destroying dead volumes in the capillary connections of your method scouting HPLC or UHPLC system by using the Thermo Scientific™ Viper™ Method Scouting Solutions kit. Viper Fingertight Fittings seal at the tip of the tubing to eliminate dead volumes and ensure easy, perfect connections in your HPLC or UHPLC system. A single kit contains all the connectors and capillaries you need and ensures consistently perfect connections.
See Viper and nanoViper resources
Specifications
Diameter (Metric) Outer100 μm
For Use With (Application)Automated Method Scouting (6 Columns)
Length (Metric)Various
MaterialMP35 N
Max. Pressure15000 psi (1034 bar)
For Use With (Equipment)Dionex UltiMate 3000 BioRS System
Product LineViper
TypeAutomated Method Scouting Solution Kit
Unit SizeEach