Product Image
Thermo Scientific™
6041RS Electrochemical Cell Replacement Gaskets
Order Thermo Scientific™ 6041RS Electrochemical Cell Replacement Gaskets when replacements are needed for your electrochemical cells.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSample Capacity
6070.252825 nL
6070.252950 nL
Catalog number 6070.2528
Price (USD)
-
Sample Capacity:
25 nL
Order Thermo Scientific™ 6041RS Electrochemical Cell Replacement Gaskets when replacements are needed for your electrochemical cells. Made of boPET, they are available in 25 and 50nL volumes.
Specifications
DescriptionQuantity: 5/Pk.
For Use With (Equipment)6041 RS Electrochemical Cell
MaterialboPET
Sample Capacity25 nL
TypeReplacement Gasket
Unit SizeEach