Vanquish™ Variable Wavelength Detector F Flow Cells
Actual product may vary
Thermo Scientific™
Vanquish™ Variable Wavelength Detector F Flow Cells
Choose the Vanquish Variable Wavelength Detector F Flow Cell based on the flow rate needed. Flow cells are available for both analytical, micro, and semi-preparative flow rates.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTypeVolume (Metric)
6077.0300Semi-Micro Bio Flow Cell2.5 μL
6077.0250Standard Flow Cell11 μL
6077.0200Standard Bio Flow Cell11 μL
6077.0360Semi-Micro Flow Cell2.5 μL
6077.0320Semi-Preparative Flow Cell0.7 μL
Catalog number 6077.0300
Price (USD)
-
Type:
Semi-Micro Bio Flow Cell
Volume (Metric):
2.5 μL
Choose the Thermo Scientific™ Vanquish™ Variable Wavelength Detector F Flow Cell based on the flow rate needed. Flow cells are available for both analytical, micro, and semi-preparative flow rates. Choose the best flow cell based on your needs for the Thermo Scientific™ Vanquish™ Variable Wavelength Detector F.
Specifications
DescriptionVanquish Variable Wavelength - Semi-Micro Bio Flow Cell
MaterialPEEK
Volume (Metric)2.5 μL
For Use With (Equipment)Vanquish Variable Wavelength Detector F
Product LineVanquish
TypeSemi-Micro Bio Flow Cell
Unit SizeEach